Learn More
Invitrogen™ MTAP Recombinant Rabbit Monoclonal Antibody (5A6V10)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA551326
Description
Immunogen sequence: MASGTTTTAV KIGIIGGTGL DDPEILEGRT EKYVDTPFGK PSDALILGKI KNVDCVLLAR HGRQHTIMPS KVNYQANIWA LKEEGCTHVI VTTACGSLRE EIQPGDIVII DQFIDRTTMR PQSFYDGSHS CARGVCHIPM AEPFCPKTRE VLIETAKKLG LRCHSKGTMV TIEGPRFSSR AESFMFRTWG ADVINMTTVP EVVLAKEAGI CYASIAMATD YDCWKEHEEA VSVDRVLKTL KENANKAKSL LLTTIPQIGS TEWSETLHNL KNMAQFSVLL PRH.
This gene encodes an enzyme that plays a major role in polyamine metabolism and is important for the salvage of both adenine and methionine. The encoded enzyme is deficient in many cancers because this gene and the tumor suppressor p16 gene are co-deleted. Multiple alternatively spliced transcript variants have been described for this gene, but their full-length natures remain unknown.
Specifications
| MTAP | |
| Recombinant Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| Q13126, Q9CQ65 | |
| MTAP | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-283 of human MTAP (NP_002442.2). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| Liquid |
| ELISA, Western Blot | |
| 5A6V10 | |
| Unconjugated | |
| MTAP | |
| 1300019I21Rik; 5'-methylthioadenosine phosphorylase; BDMF; c86fus; Cc1-6; DMSFH; DMSMFH; epididymis luminal protein 249; HEL-249; LGMBF; MeSAdo phosphorylase; methylthioadenosine phosphorylase; MSAP; MTA phosphorylase; MTAP; MTAPase; S-methyl-5'-thioadenosine phosphorylase; zgc:66012 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 298227, 4507, 66902 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.