missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTAP Polyclonal Antibody

MTAP Polyclonal Antibody
Brand: Thermo Scientific PA5106861
This item is not returnable.
View return policy
Description
MTAP Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
Specifications
| MTAP | |
| Polyclonal | |
| Unconjugated | |
| MTAP | |
| 1300019I21Rik; 5'-methylthioadenosine phosphorylase; BDMF; c86fus; Cc1-6; DMSFH; DMSMFH; epididymis luminal protein 249; HEL-249; LGMBF; MeSAdo phosphorylase; methylthioadenosine phosphorylase; MSAP; MTA phosphorylase; MTAP; MTAPase; S-methyl-5'-thioadenosine phosphorylase; zgc:66012 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 298227, 4507, 66902 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q13126, Q9CQ65 | |
| MTAP | |
| A synthesized peptide derived from human MTAP(Accession Q13126), corresponding to amino acid residues L204-T254. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction