missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTAP Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
£161.00 - £383.00
Specifications
| Antigen | MTAP |
|---|---|
| Dilution | Western Blot 1:1000 - 1:5000, ELISA Recommended starting concentration is 1 μg/mL |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30232874
|
Novus Biologicals
NBP3-33318-20ul |
20 μL |
£161.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30232522
|
Novus Biologicals
NBP3-33318-100ul |
100 μL |
£383.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MTAP Monoclonal antibody specifically detects MTAP in Human,Mouse,Rat samples. It is validated for ELISA,Western BlotSpecifications
| MTAP | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| 5'-methylthioadenosine phosphorylase, c86fus, EC 2.4.2.28, methylthioadenosine phosphorylase, MSAPMeSAdo phosphorylase, MTA phosphorylase, MTAPase, S-methyl-5'-thioadenosine phosphorylase | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-283 of human MTAP (NP_002442.2).,, Sequence:, MASGTTTTAVKIGIIGGTGLDDPEILEGRTEKYVDTPFGKPSDALILGKIKNVDCVLLARHGRQHTIMPSKVNYQANIWALKEEGCTHVIVTTACGSLREEIQPGDIVIIDQFIDRTTMRPQSFYDGSHSCARGVCHIPMAEPFCPKTREVLIETAKKLGLRCHSKGTMVTIEGPRFSSRAESFMFRTWGADVINMTTVPEVVLAKEAGICYASIAMATDYDCWKEHEEAVSVDRVLKTLKENANKAKSLLLTTIPQIGSTEWSETLHNLKNMAQFSVLLPRH | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:1000 - 1:5000, ELISA Recommended starting concentration is 1 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| 4507 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title