missing translation for 'onlineSavingsMsg'
Learn More

MTAP Antibody (MTAP/1813), Novus Biologicals™

Mouse Monoclonal Antibody

£236.00 - £392.00

Specifications

Antigen MTAP
Clone MTAP/1813
Concentration 0.2 mg/mL
Dilution Western Blot 1-2 μg/mL, ELISA 2-4 μg/mL, Immunohistochemistry, Immunohistochemistry-Paraffin 1-2 μg/mL, Protein Array
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18651758
View Documents
Novus Biologicals
NBP2-75730-100ug
100 μg
£392.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18643719
View Documents
Novus Biologicals
NBP2-75730-20ug
20 μg
£236.00
20µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

MTAP Monoclonal antibody specifically detects MTAP in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array
Specifications

Specifications

MTAP
0.2 mg/mL
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array
Unconjugated
Mouse
Human
Q13126
4507
IgG2b κ
Protein A or G purified
MTAP/1813
Western Blot 1-2 μg/mL, ELISA 2-4 μg/mL, Immunohistochemistry, Immunohistochemistry-Paraffin 1-2 μg/mL, Protein Array
Monoclonal
Purified
RUO
10 mM PBS with 0.05% BSA
5'-methylthioadenosine phosphorylase, c86fus, EC 2.4.2.28, methylthioadenosine phosphorylase, MSAPMeSAdo phosphorylase, MTA phosphorylase, MTAPase, S-methyl-5'-thioadenosine phosphorylase
Recombinant human MTAP protein fragment (aa97-196) (exact sequence is proprietary) (Uniprot: Q13126)
Primary
Store at 4°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.