missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTAP Antibody (JE63-74), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-32610
This item is not returnable.
View return policy
Description
MTAP Monoclonal antibody specifically detects MTAP in Human,Mouse,Rat samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/Immunofluorescence
Specifications
| MTAP | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000, Flow Cytometry 1:1000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:500 | |
| 5'-methylthioadenosine phosphorylase, c86fus, EC 2.4.2.28, methylthioadenosine phosphorylase, MSAPMeSAdo phosphorylase, MTA phosphorylase, MTAPase, S-methyl-5'-thioadenosine phosphorylase | |
| Recombinant protein within Human MTAP aa 184-283/ 283. (Uniprot: Q13126) | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Flow Cytometry, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| JE63-74 | |
| Unconjugated | |
| 1*TBS (pH7.4), 0.05% BSA and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 4507 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction