Learn More
Invitrogen™ MTA3 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA540670
Product Code. 15544322
  • £441.00 / 100µL
Description
Peptide sequence: TLIMLADKHA KEIEEESETT VEADLTDKQK HQLKHRELFL SRQYESLPAT Sequence homology: Cow: 100%; Dog: 100%; Horse: 100%; Human: 100%; Mouse: 92%; Rabbit: 100%; Rat: 100%.
Plays a role in maintenance of the normal epithelial architecture through the repression of SNAI1 transcription in a histone deacetylase-dependent manner, and thus the regulation of E-cadherin levels. Contributes to transcriptional repression by BCL6. [UniProt].
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MTA3
Polyclonal
Unconjugated
Mta3
1110002J22Rik; KIAA1266; metastasis associated 1 family member 3; metastasis associated 1 family, member 3; metastasis associated 3; metastasis associated family, member 3; metastasis associated gene family, member 3; metastasis-associated protein MTA3; mKIAA1266; Mta3
Rabbit
Affinity Chromatography
RUO
57504
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
0.5 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
Q9BTC8
Mta3
Synthetic peptide directed towards the N-terminal of human MTA3 (aa 54-103).