missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTA3 Monoclonal Antibody (3B3), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA522371
This item is not returnable.
View return policy
Description
Peptide Sequence: LKMPTQSEEE KLSPSPTTED PRVRSHVSRQ AMQGMPVRNT GSPKSAVKTR QAFFLHTTYF TKFARQVCKN TLRLRQAARR PFVAINYAAI RAECKMLLNS
Plays a role in maintece of the normal epithelial architecture through the repression of SNAI1 transcription in a histone deacetylase-dependent manner, and thus the regulation of E-cadherin levels.
Specifications
| MTA3 | |
| Monoclonal | |
| 1.0 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Q9BTC8 | |
| MTA3 | |
| MTA3 (NP_065795, 416 a.a. ∽ 515 a.a) partial recombit protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b κ |
| ELISA, Immunocytochemistry, Immunofluorescence | |
| 3B3 | |
| Unconjugated | |
| MTA3 | |
| metastasis associated family, member 3, metastasis associated gene family, member 3, metastasis-associated protein MTA3 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 57504 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction