missing translation for 'onlineSavingsMsg'
Learn More

MTA3 Antibody (OTI2B8), Novus Biologicals Biologicals™

Mouse Monoclonal Antibody

£195.00 - £417.00

Specifications

Antigen MTA3
Clone OTI2B8
Dilution Western Blot 1:500-2000
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30306435
View Documents
Novus Biologicals
NBP3-45015-100ul
100 μL
£417.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30306719
View Documents
Novus Biologicals
NBP3-45015-30ul
30 μL
£195.00
30µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

MTA3 Monoclonal antibody specifically detects MTA3 in Human,Mouse,Rat samples. It is validated for Western Blot
Specifications

Specifications

MTA3
Western Blot 1:500-2000
Monoclonal
Purified
RUO
Human, Mouse, Rat
member 3, metastasis associated 1 family, member 3, metastasis-associated protein MTA3
Human recombinant protein fragment corresponding to amino acids 1-259 of human MTA3 (NP_065795) produced in E.coli.
Primary
Store at -20 degrees C. Avoid freeze/thaw cycles.
OTI2B8
Western Blot
Unconjugated
Mouse
Breast Cancer, Cancer
PBS (pH 7.3) 1% BSA, 50% glycerol
57504
IgG1
>95%
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.