missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTA3 Antibody (2C5), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00057504-M06
This item is not returnable.
View return policy
Description
MTA3 Monoclonal antibody specifically detects MTA3 in Human samples. It is validated for ELISA, Immunocytochemistry/ Immunofluorescence
Specifications
| MTA3 | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| member 3, metastasis associated 1 family, member 3, metastasis-associated protein MTA3 | |
| MTA3 (NP_065795, 416 a.a. ∽ 515 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. LKMPTQSEEEKLSPSPTTEDPRVRSHVSRQAMQGMPVRNTGSPKSAVKTRQAFFLHTTYFTKFARQVCKNTLRLRQAARRPFVAINYAAIRAECKMLLNS | |
| 0.1 mg | |
| Breast Cancer, Cancer | |
| 57504 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG2b κ |
| ELISA, Immunofluorescence | |
| 2C5 | |
| ELISA, Immunocytochemistry/ Immunofluorescence | |
| NP_065795 | |
| Mouse | |
| IgG purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction