missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LGR6 Recombinant Rabbit Monoclonal Antibody (HL2827)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556779
This item is not returnable.
View return policy
Description
LGR6 Recombinant Monoclonal Antibody for Western Blot, IHC (P)
LGR6 has been reported to be expressed in rat adrenal, brain, colon, heart, kidney, ovary, oviduct, small intestine, spleen, testis, and uterus. ESTs have been isolated from human B-cell/lung/testis, blood, brain, colon, eye, germ cell, head/neck, heart, heart/melanocyte/uterus, lung, ovary, stomach, testis, and uterus libraries.
Specifications
| LGR6 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q9HBX8 | |
| Lgr6 | |
| Recombinant protein encompassing a sequence within the Intracellular domain of human LGR6. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| HL2827 | |
| Unconjugated | |
| Lgr6 | |
| A530037C04Rik; D830026M09; FLJ14471; gonadotropin receptor; GPCR; leucine rich repeat containing G protein-coupled receptor 6; leucine-rich repeat containing G protein-coupled receptor 6; leucine-rich repeat-containing G protein-coupled receptor 6; leucine-rich repeat-containing G-protein coupled receptor 6; Lgr6; RGD1560339; RP11-572A16.3; UNQ6427/PRO21331; VTS20631 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 59352 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction