missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MTA2 Polyclonal Antibody

Description
In Western blot applications, this antibody detects a band at ~75kDa. A suggested positive control is Hela cell lysate. PA5-19978 can be used with blocking peptide PEP-0102.
This gene encodes a protein that has been identified as a component of NuRD, a nucleosome remodeling deacetylase complex identified in the nucleus of human cells. It shows a very broad expression pattern and is strongly expressed in many tissues. It may represent one member of a small gene family that encode different but related proteins involved either directly or indirectly in transcriptional regulation. Their indirect effects on transcriptional regulation may include chromatin remodeling. It is closely related to another member of this family, a protein that has been correlated with the metastatic potential of certain carcinomas. These two proteins are so closely related that they share the same types of domains. These domains include two DNA binding domains, a dimerization domain, and a domain commonly found in proteins that methylate DNA. One of the proteins known to be a target protein for this gene product is p53. Deacetylation of p53 is correlated with a loss of growth inhibition in transformed cells supporting a connection between these gene family members and metastasis.
Specifications
Specifications
| Antigen | MTA2 |
| Applications | Immunohistochemistry, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | Mta2 |
| Gene Accession No. | O94776, Q9R190 |
| Gene Alias | AW550797; DKFZp686F2281; Mata1l1; metastais-associated gene family, member 2; metastasis associated 1 family member 2; metastasis associated 1 family, member 2; metastasis associated 1-like 1; metastasis -associated gene 1-like 1; metastasis associated gene family, member 2; metastasis-associated 1-like 1; metastasis-associated gene family, member 2; metastasis-associated protein 2; metastasis-associated protein MTA2; mmta2; Mta1l1; MTA1-L1 protein; Mta2; p53 target protein in deacetylase complex; PID |
| Gene Symbols | Mta2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?