missing translation for 'onlineSavingsMsg'
Learn More

MTA2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 2 publications

Brand:  Novus Biologicals NB100-56483SS

 View more versions of this product

Product Code. 18038155

  • £167.00 / 25µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

MTA2 Polyclonal specifically detects MTA2 in Human, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MTA2
Polyclonal
Unconjugated
PBS containing 0.05% BSA with 0.05% Sodium Azide
DKFZp686F2281, metastasis associated 1 family, member 2, metastasis -associated gene 1-like 1, metastasis associated gene family, member 2, Metastasis-associated 1-like 1, metastasis-associated protein 2, metastasis-associated protein MTA2, MTA1-L1 protein, MTA1L1MTA1-L1, p53 target protein in deacetylase complex, PID
Rabbit
75 kDa
0.025 mg
Cancer, Chromatin Research, Transcription Factors and Regulators, Tumor Suppressors
9219
Human, Primate
IgG
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
0.5 mg/mL
Western Blot 0.5-2 ug/ml, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:1000, Immunohistochemistry-Paraffin 5-10 ug/ml
O94776
MTA2
A portion of amino acids 650-700 of human MTA2 was used as the immunogen (NP_004730).
Affinity Purified
RUO
Primary
Reacts with Chimpanzee, Human and Rhesus Monkey. Predicted cross-reactivity based on sequence identity: Mouse (94%), Rat (94%), Dog (88%), Bovine (88%) and Xenopus (76%).
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only