missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTA2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93800-0.1ml
This item is not returnable.
View return policy
Description
MTA2 Polyclonal antibody specifically detects MTA2 in Human, Mouse, Rat samples. It is validated for Western Blot, ChIP assay (ChIP), Gene Knock-Out
Specifications
| MTA2 | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000, Chromatin Immunoprecipitation (ChIP) 1:50-1:200, Knockout Validated | |
| DKFZp686F2281, metastasis associated 1 family, member 2, metastasis -associated gene 1-like 1, metastasis associated gene family, member 2, Metastasis-associated 1-like 1, metastasis-associated protein 2, metastasis-associated protein MTA2, MTA1-L1 protein, MTA1L1MTA1-L1, p53 target protein in deacetylase complex, PID | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 60-180 of human MTA2 (NP_004730.2). DSNAREFEEESKQPGVSEQQRHQLKHRELFLSRQFESLPATHIRGKCSVTLLNETDILSQYLEKEDCFFYSLVFDPVQKTLLADQGEIRVGCKYQAEIPDRLVEGESDNRNQQKMEMKVWD | |
| 0.1 mL | |
| Cancer, Chromatin Research, Transcription Factors and Regulators, Tumor Suppressors | |
| 9219 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ChIP Assay, Gene Knock-Out | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction