missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
MTA2 Polyclonal antibody specifically detects MTA2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | MTA2 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 594 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | DKFZp686F2281, metastasis associated 1 family, member 2, metastasis -associated gene 1-like 1, metastasis associated gene family, member 2, Metastasis-associated 1-like 1, metastasis-associated protein 2, metastasis-associated protein MTA2, MTA1-L1 protein, MTA1L1MTA1-L1, p53 target protein in deacetylase complex, PID |
| Host Species | Rabbit |
| Immunogen | Antibody was raised against a 17 amino acid peptide near the carboxy terminus of human PID. The immunogen is located within the last 50 amino acids of PID. Amino Acid Squence: PAPSHPASTNEPIVLED |
| Purification Method | Peptide affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?