missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTA1 Monoclonal Antibody (1F3F12), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen 390700
This item is not returnable.
View return policy
Description
This antibody is specific for the C-terminal region of the human MTA1 protein. On Western blots, it identifies doublets at ∽80 kDa. A band of unknown origin is observed at ∽150 kDa.
This gene encodes a protein that was identified in a screen for genes expressed in metastatic cells, specifically, mammary adenocarcinoma cell lines. Expression of this gene has been correlated with the metastatic potential of at least two types of carcinomas although it is also expressed in many normal tissues. The role it plays in metastasis is unclear. It was initially thought to be the 70kD component of a nucleosome remodeling deacetylase complex, NuRD, but it is more likely that this component is a different but very similar protein. These two proteins are so closely related, though, that they share the same types of domains. These domains include two DNA binding domains, a dimerization domain, and a domain commonly found in proteins that methylate DNA. The profile and activity of this gene product suggest that it is involved in regulating transcription and that this may be accomplished by chromatin remodeling.
Specifications
| MTA1 | |
| Monoclonal | |
| Conc. Not Determined | |
| tissue culture supernatant with 0.1% sodium azide | |
| Q13330, Q62599, Q8K4B0 | |
| MTA1 | |
| Synthetic peptide derived from the N-terminal region of the human MST3 protein | |
| RUO | |
| 116870, 64520, 9112 | |
| Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 1F3F12 | |
| Unconjugated | |
| MTA1 | |
| metastasis associated 1; metastasis associated gene 1 protein; metastasis-associated protein MTA1; MTA1; Zg29 | |
| Mouse | |
| 1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction