missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ MTA1 Antibody [PerCP], Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals™ NB600-260PCP

Product Code. 30025617

  • £467.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

MTA1 Polyclonal antibody specifically detects MTA1 in Human,Mouse samples. It is validated for Immunohistochemistry,Immunoprecipitation,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MTA1
Polyclonal
PBS
Rabbit
Immunogen affinity purified
RUO
Primary
Human, Mouse
Purified
Immunohistochemistry, Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin)
PerCP
metastasis associated 1, metastasis associated gene 1 protein, metastasis associated protein, metastasis-associated protein MTA1
The immunogen recognized by this antibody maps to a region between residues 400 and 450 of human Metastasis Associated Protein 1 using the numbering given in Swiss-Prot entry Q13330 (GeneID 9112).
0.1 mL
Cancer, Cellular Markers, Chromatin Research, Transcription Factors and Regulators
9112
Store at 4°C in the dark.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.