missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MT3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12179-1-AP-150UL
This item is not returnable.
View return policy
Description
Metallothionein (MT) is a sulfhydryl- and cysteine-rich protein found in micro-organisms, plants and all invertebrate and vertebrate animals. Metallothioneins are a group of ubiquitous low-molecular-weight proteins that have functional roles in cell growth, repair and differentiation. Metallothionein are implicated primarily in metal ion detoxification as they are essential for the protection of cells against the toxicity of cadmium, mercury and copper. Metallothioneins are known to be broadly expressed in heart, liver, kidney, breast and testis tissue. Metallothionein 3, also known as MT-3 or GIFB (growth inhibitory factor), is a 68 amino acid protein that belongs to the type 1 family and Metallothionein superfamily. While highly expressed in astrocytes of the normal human brain, Metallothionein 3 expression is reduced in the brains of patients with Alzheimer disease.Specifications
| MT3 | |
| Polyclonal | |
| Unconjugated | |
| MT3 | |
| GIF, GIFB, GRIF, Growth inhibitory factor, metallothionein 3, Metallothionein III, MT 3, MT III, MT3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 117038, 17751, 4504 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P25713, P28184, P37361 | |
| Mt3 | |
| MT3 Fusion Protein Ag2823 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction