missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MT1A, Rabbit anti-Human, Clone: N11-G, Abnova™
Rabbit Monoclonal Antibody
Brand: Abnova MAB15897.100uL
This item is not returnable.
View return policy
Specifications
| MT1A | |
| Monoclonal | |
| Unconjugated | |
| In 20 mM Tris-HCl buffer, pH 8.0 (10 mg/mL BSA, 0.05% Sodium Azide). | |
| MGC32848/MT1/MT1S/MTC | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Human, Rabbit | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| N11-G | |
| ELISA (1:5000-1:10000) Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| metallothionein 1A | |
| MT1A | |
| A synthetic peptide corresponding to N-terminus of human Metallothionein. | |
| RUO | |
| 4489/4490/4493/4494/4495/4496/4499/4490/4489/4502 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction