missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MT-ND6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA543532
This item is not returnable.
View return policy
Description
Peptide sequence: DGVVVVVNFN SVGSWMIYEG EGSGLIREDP IGAGALYDYG RWLVVVTGWT Sequence homology: Human: 100%; Pig: 93%.
Core subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I) that is believed to belong to the minimal assembly required for catalysis. Complex I functions in the transfer of electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone.
Specifications
| MT-ND6 | |
| Polyclonal | |
| Unconjugated | |
| mt-Nd6 | |
| mitochondrially encoded NADH dehydrogenase 6; MTND6; MT-ND6; NADH dehydrogenase subunit 6; NADH6; NADH-ubiquinone oxidoreductase chain 6; ND6 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 4541 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot, Surface Plasmon Resonance | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P03923 | |
| mt-Nd6 | |
| Synthetic peptide directed towards the middle region of human ND6. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction