missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MT-ND6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA550710
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total MT-ND6 protein.
Core subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I) that is believed to belong to the minimal assembly required for catalysis. Complex I functions in the transfer of electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone.
Specifications
| MT-ND6 | |
| Polyclonal | |
| Unconjugated | |
| mt-Nd6 | |
| mitochondrially encoded NADH dehydrogenase 6; MTND6; MT-ND6; NADH dehydrogenase subunit 6; NADH6; NADH-ubiquinone oxidoreductase chain 6; ND6 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 4541 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 2.4 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| P03923 | |
| mt-Nd6 | |
| Synthetic peptide corresponding to a region derived from internal residues of human mitochondrially encoded NADH dehydrogenase 6. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction