missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MT-ND5 Polyclonal Antibody

MT-ND5 Polyclonal Antibody
Brand: Invitrogen PA5107191
This item is not returnable.
View return policy
Description
MT-ND5 Polyclonal Antibody for Western Blot
Specifications
| MT-ND5 | |
| Polyclonal | |
| Unconjugated | |
| MT-ND5 | |
| mitochondrially encoded NADH dehydrogenase 5; MTND5; mt-Nd5; NADH Dehydrogenase 5; NADH dehydrogenase subunit 5; NADH5; NADH-ubiquinone oxidoreductase chain 5; ND5 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 4540 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 | |
| P03915 | |
| MT-ND5 | |
| Synthetic peptide directed towards the N terminal region of human ND5. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction