missing translation for 'onlineSavingsMsg'
Learn More

MT-ND4L Antibody, Novus Biologicals™

Product Code. 18360172 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18360172 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18360172 Supplier Novus Biologicals Supplier No. NBP305239

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MT-ND4L Polyclonal specifically detects MT-ND4L in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MT-ND4L
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias Complex I ND4L Subunit, EC 1.6.5.3, Mitochondrially Encoded NADH Dehydrogenase 4L, Mitochondrially Encoded NADH:Ubiquinone, MTND4L, NADH Dehydrogenase 4L, NADH Dehydrogenase, Subunit 4L (Complex I), NADH4L, NADH-Ubiquinone Oxidoreductase Chain 4L, ND4L, Oxidoreductase Core Subunit 4L
Host Species Rabbit
Immunogen Recombinant protein of human MT-ND4L.
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4539
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.