missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MT-ND3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£222.00 - £513.00
Specifications
| Antigen | MT-ND3 |
|---|---|
| Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18627315
|
Novus Biologicals
NBP2-38021-25ul |
25 μL |
£222.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18114339
|
Novus Biologicals
NBP2-38021 |
0.1 mL |
£513.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MT-ND3 Polyclonal specifically detects MT-ND3 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| MT-ND3 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| P03897 | |
| 4537 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: GYIEKSTPYECGFDPISPARVPFSIK | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| Complex I ND3 Subunit, EC 1.6.5.3, Mitochondrially Encoded NADH Dehydrogenase 3, MTND3, NADH Dehydrogenase 3, NADH Dehydrogenase Subunit 3, NADH Dehydrogenase, Subunit 3 (Complex I), NADH3, NADH-Ubiquinone Oxidoreductase Chain 3, ND3 | |
| ND3 | |
| IgG | |
| Affinity Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title