missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MT-ND1 Antibody (JE34-93), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-32613
This item is not returnable.
View return policy
Description
MT-ND1 Monoclonal antibody specifically detects MT-ND1 in Human,Mouse,Rat samples. It is validated for Western Blot
Specifications
| MT-ND1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000 | |
| EC 1.6.5.3, mitochondrially encoded NADH dehydrogenase 1, MTND1, NAD1, NADH dehydrogenase subunit 1, NADH dehydrogenase, subunit 1 (complex I), NADH1, ND1NADH dehydrogenase 1 | |
| Synthetic peptide within Human MT-ND1 aa 23-67/ 318. (Uniprot: P03886) | |
| 100 μL | |
| Cancer, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction | |
| 4535 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| JE34-93 | |
| Unconjugated | |
| PBS (pH7.4), 0.05% BSA and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction