missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MT-CYB Polyclonal Antibody
GREENER_CHOICE

Product Code. 17161784 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17161784 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17161784 Supplier Invitrogen™ Supplier No. PA5113926

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: LFSPDLLGDP DNYTLANPLN TPPHIKPEWY FLFAYTILRS VPNKLGGVLA For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Human: 100%.

CYTB belongs to the cytochrome b family. It is a component of the ubiquinol-cytochrome c reductase complex (complex III or cytochrome b-c1 complex), which is a respiratory chain that generates an electrochemical potential coupled to ATP synthesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MT-CYB
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene MT-CYB
Gene Accession No. P00156
Gene Alias COB; Complex III subunit 3; Complex III subunit III; CYTB; Cytochrome b; Cytochrome b-c1 complex subunit 3; mitochondrially encoded cytochrome b; Mtcyb; Mt-Cyb; mt-Cytb; Ubiquinol-cytochrome-c reductase complex cytochrome b subunit; UQCR3
Gene Symbols MT-CYB
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal region of human MT-CYB (aa 244-293).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4519
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.