missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MT-ATP6 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA5116789
This item is not returnable.
View return policy
Description
Positive Samples: HeLa, U-251MG, mouse brain, mouse heart, mouse kidney, rat kidney Immunogen sequence: PPLLIPTSKY LINNR
The MT-ATP6 gene provides information for making a protein that is essential for normal mitochondrial function. Mitochondria are structures within cells that convert the energy from food into a form that cells can use. These cellular structures produce energy through a process called oxidative phosphorylation, which uses oxygen and simple sugars to create adenosine triphosphate (ATP), the cell's main energy source. The MT-ATP6 protein forms one part (subunit) of a large enzyme called ATP synthase. This enzyme, which is also known as complex V, is responsible for the final step of oxidative phosphorylation. Specifically, one segment of ATP synthase allows positively charged particles, called protons, to flow across a specialized membrane inside mitochondria. Another segment of the enzyme uses the energy created by this proton flow to convert a molecule called adenosine diphosphate (ADP) to ATP.
Specifications
| MT-ATP6 | |
| Polyclonal | |
| Unconjugated | |
| MT-ATP6 | |
| ATP synthase F0 subunit 6; ATP synthase subunit a; Atp6; ATPase6; F-ATPase protein 6; mitochondrially encoded ATP synthase 6; Mtatp6; MT-ATP6 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 17705, 26197, 4508 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| 2.14 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| P00846, P00848, P05504 | |
| MT-ATP6 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human MT-ATP6 (YP_0030240311) | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction