missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MSY2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | MSY2 |
|---|---|
| Concentration | 0.33 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
YBX2 is a germ-cell specific member of the vertebrate Y box family of nucleic acid-binding proteins.Specifications
| MSY2 | |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Rat, Human, Mouse | |
| Q9Y2T7, Q9Z2C8 | |
| 303250, 51087, 53422 | |
| MSY2 Fusion Protein Ag4465 | |
| Primary | |
| -20°C | |
| YBX2 |
| 0.33 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| CONTRIN, CSDA3, DBPC, DNA binding protein C, MSY2, MSY2 homolog, Y box binding protein 2, YBX2 | |
| Ybx2 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title