missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MSY2 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£143.00 - £274.00
Specifications
| Antigen | MSY2 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18345556
|
Novus Biologicals
NBP3-16007-20UL |
20 μg |
£143.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18396416
|
Novus Biologicals
NBP3-16007-100UL |
100 μg |
£274.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MSY2 Polyclonal antibody specifically detects MSY2 in Human, Mouse samples. It is validated for Western BlotSpecifications
| MSY2 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS, 50% glycerol, pH7.3 | |
| 51087 | |
| IgG | |
| Affinity purified |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| Contrin, CSDA3MGC45104, DBPC, DNA-binding protein C, Germ cell-specific Y-box-binding protein, MSY2 homolog, MSY2germ cell specific Y-box binding protein, Y box binding protein 2, Y-box-binding protein 2 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human MSY2 (NP_057066.2). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title