missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Msx3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA568815
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Mouse: 100%; Rat: 100%.
Involved in the development of the inner ear.
Specifications
| Msx3 | |
| Polyclonal | |
| Unconjugated | |
| Msx3 | |
| AI323377; etID309953.13; homeo box, msh-like 3; homeobox; homeobox protein MSH-C; homeobox protein MSX-3; homeobox, msh-like 3; msh homeobox 3; mshC; msh-C; Msx3; Msx-3; msxc; msx-c; muscle segment homeobox 3; muscle segment homeobox C; zgc:86596 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 114504 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| 0 | |
| Msx3 | |
| synthetic peptide directed towards the C-terminal region of rat Msx3. | |
| 100 μL | |
| Primary | |
| Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction