Learn More
MSX2 Mouse, anti-Human, Clone: 1B1G3, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68550-1-IG-150UL
This item is not returnable.
View return policy
Description
The MSX2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human MSX2. This antibody recognizes human antigen. The MSX2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| MSX2 | |
| 1B1G3 | |
| CRS2, FPP, Homeobox protein Hox 8, Homeobox protein MSX 2, HOX8, MSH, msh homeobox 2, MSX2, PFM, PFM1 | |
| 150 μL | |
| 4488 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
| Western Blot, ELISA | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG1 |