missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MSX1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£292.00 - £460.00
Specifications
| Antigen | MSX1 |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18229813
|
Novus Biologicals
NBP2-57021 |
100 μL |
£460.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18623488
|
Novus Biologicals
NBP2-57021-25ul |
25 μL |
£292.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MSX1 Polyclonal specifically detects MSX1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| MSX1 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| Homeobox protein Hox-7, homeobox protein MSX-1, HOX7homeobox 7, HYD1msh homeobox homolog 1 (Drosophila), msh (Drosophila) homeo box homolog 1 (formerly homeo box 7), msh homeo box 1, msh homeobox 1, Msh homeobox 1-like protein, msh homeobox homolog 1, OFC5, STHAG1 | |
| MSX1 | |
| IgG | |
| Affinity Purified |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 4487 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:MAPAADMTSLPLGVKVEDSAFGKP | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title