missing translation for 'onlineSavingsMsg'
Learn More

MSX-122, MedChemExpress

Product Code. p-200101986
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16488477

Brand: Medchem Express HY13696

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress MSX-122 is an orally active partial antagonist of CXCR4, inhibiting CXCR4/CXCL12 actions, with an IC50 of ∼10 nM. MSX-122 has anti-inflammatory and anti-metastatic activity.

MSX-122 is an orally active partial antagonist of CXCR4, inhibiting CXCR4/CXCL12 actions, with an IC50 of ∼10 nM. MSX-122 has anti-inflammatory and anti-metastatic activity.

Specifications

Chemical Name or Material MSX-122
Physical Form Solid
CAS 897657-95-3
Color Yellow
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C16H16N6
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : 4 mg/mL (13.68 mM; Need ultrasonic)
SMILES C1(CNC2=NC=CC=N2)=CC=C(CNC3=NC=CC=N3)C=C1
Molecular Weight (g/mol) 292.34
Formula Weight 292.34
Percent Purity 96.56%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.