Learn More
Invitrogen™ Human MST4, GST Tag Recombinant Protein
Description
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Full Length
Specifications
Specifications
| Accession Number | Q9P289 |
| Concentration | See Label |
| For Use With (Application) | Kinase Assay |
| Formulation | 50 mM tris with 0.02% Triton X-100, 0.5 mM EDTA, 150 mM NaCl, 2 mM DTT, 50% glycerol and no preservative; pH 7.5 |
| Gene ID (Entrez) | 51765 |
| Molecular Weight (g/mol) | 75.7 kDa |
| Name | Human MST4, GST Tag |
| Quantity | 100 μg |
| Regulatory Status | RUO - research use only |
| Gene Alias | 2610018G03Rik; Mammalian STE20-like protein kinase 4; mammalian sterile 20-like 4; Mask; Mst3 and SOK1-related kinase; Mst4; MST-4; OTTHUMP00000215132; OTTHUMP00000215134; RGD1563568; RP6-213H19.1; serine/threonine kinase 26; serine/threonine protein kinase 26; serine/threonine protein kinase MST4; serine/threonine-protein kinase 26; Serine/threonine-protein kinase MASK; serine/threonine-protein kinase MST4; STE20-like kinase 4; STE20-like kinase MST4; STK26 |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.