missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MST4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-1584
This item is not returnable.
View return policy
Description
MST4 Polyclonal specifically detects MST4 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| MST4 | |
| Polyclonal | |
| Unconjugated | |
| TBS and 0.1% BSA with 0.09% Sodium Azide | |
| EC 2.7.11, EC 2.7.11.1, Mammalian STE20-like protein kinase 4, mammalian sterile 20-like 4, MASK, Mst3 and SOK1-related kinase, MST-4, serine/threonine protein kinase MASK, serine/threonine protein kinase MST4, Serine/threonine-protein kinase MASK, serine/threonine-protein kinase MST4, STE20-like kinase MST4 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG |
| Western Blot, Immunoprecipitation | |
| 0.2 mg/mL | |
| Western Blot 1:1000-1:10000, Immunoprecipitation | |
| NP_057626.2 | |
| MST4 | |
| The immunogen recognized by this antibody maps to a region between residue 375 and the C-terminus (residue 416) of human Mammalian Ste20-like Kinase 4 (Mst3 and SOK1-related kinase) using the numbering given in entry NP_057626.2 (GeneID 51765). | |
| 0.1 mL | |
| Protein Kinase | |
| 51765 | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction