missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MST4 Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
£161.00 - £383.00
Specifications
| Antigen | MST4 |
|---|---|
| Dilution | Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30231517
|
Novus Biologicals
NBP3-33253-20ul |
20 μL |
£161.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30232295
|
Novus Biologicals
NBP3-33253-100ul |
100 μL |
£383.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MST4 Monoclonal antibody specifically detects MST4 in Human,Mouse,Rat samples. It is validated for ELISA,Western BlotSpecifications
| MST4 | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Protein Kinase | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| 51765 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| EC 2.7.11, EC 2.7.11.1, Mammalian STE20-like protein kinase 4, mammalian sterile 20-like 4, MASK, Mst3 and SOK1-related kinase, MST-4, serine/threonine protein kinase MASK, serine/threonine protein kinase MST4, Serine/threonine-protein kinase MASK, serine/threonine-protein kinase MST4, STE20-like kinase MST4 | |
| A synthetic peptide corresponding to a sequence within amino acids 317-416 of human MST4 (Q9P289).,, Sequence:, ENNTHPEWSFTTVRKKPDPKKVQNGAEQDLVQTLSCLSMIITPAFAELKQQDENNASRNQAIEELEKSIAVAEAACPGITDKMVKKLIEKFQKCSADESP | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title