missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MST4 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-1583CL1
This item is not returnable.
View return policy
Description
MST4 Polyclonal antibody specifically detects MST4 in Human samples. It is validated for Immunoprecipitation,Western Blot (Negative)
Specifications
| MST4 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation, Western Blot (Negative) | |
| CoraFluor 1 | |
| EC 2.7.11, EC 2.7.11.1, Mammalian STE20-like protein kinase 4, mammalian sterile 20-like 4, MASK, Mst3 and SOK1-related kinase, MST-4, serine/threonine protein kinase MASK, serine/threonine protein kinase MST4, Serine/threonine-protein kinase MASK, serine/threonine-protein kinase MST4, STE20-like kinase MST4 | |
| The immunogen recognized by this antibody maps to a region between residues 300 and 350 of human Mammalian Ste20-like Kinase 4 (Mst3 and SOK1-related kinase) using the numbering given in entry NP_057626.2 (GeneID 51765). | |
| 0.1 mL | |
| Protein Kinase | |
| 51765 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction