Learn More
Invitrogen™ MST3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA522269
Description
Recommended positive controls: 293T, A431, H1299, HeLa, Raji, rat brain. Predicted reactivity: Mouse (100%), Rat (100%), Xenopus laevis (100%), Chicken (100%), Rhesus Monkey (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
STK24 (MST3) is a serine/threonine kinase that is a member of the GCK subfamily of STE20-like proteins. The yeast 'Sterile 20' gene (STE20) functions upstream of the mitogen-activated protein kinase (MAPK) cascade. In mammals, protein kinases related to STE20 can be divided into 2 subfamilies based on their structure and regulation. Members of the PAK subfamily (see PAK3; MIM 300142) contain a C-terminal catalytic domain and an N-terminal regulatory domain that has a CDC42 (MIM 116952)-binding domain. In contrast, members of the GCK subfamily (see MAP4K2; MIM 603166), also called the Sps1 subfamily, have an N-terminal catalytic domain and a C-terminal regulatory domain without a CDC42-binding domain. STK24 belongs to the GCK subfamily of STE20-like kinases.
Specifications
| MST3 | |
| Polyclonal | |
| Unconjugated | |
| STK24 | |
| 1810013H02Rik; C76483; epididymis secretory protein Li 95; HEL-S-95; Mammalian STE20-like protein kinase 3; Mammalian STE20-like protein kinase 3 C-terminal; Mammalian STE20-like protein kinase 3 N-terminal; mammalian sterile twenty 3 kinase; Mst3; MST-3; MST3/C; MST3/N; MST3B; OTTHUMP00000018594; RGD1561742; RP11-111L24.5; serine/threonine kinase 24; serine/threonine kinase 24 (STE20 homolog, yeast); serine/threonine-protein kinase 24; Serine/threonine-protein kinase 24 12 kDa subunit; Serine/threonine-protein kinase 24 35 kDa subunit; Serine/threonine-protein kinase 24 36 kDa subunit; STE20; STE20-like kinase 3; STE20-like kinase MST3; sterile 20-like kinase 3; STK24; STK3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 361092, 8428 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.94 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| B0LT89, Q9Y6E0 | |
| STK24 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the center region of human MST3. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.