missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
MST3 Monoclonal antibody specifically detects MST3 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Specifications
Specifications
| Antigen | MST3 |
| Applications | Western Blot, ELISA, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 3F2 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500, ELISA, Immunocytochemistry/ Immunofluorescence |
| Formulation | In 1x PBS, pH 7.4 |
| Gene Accession No. | AAH35578 |
| Gene Alias | EC 2.7.11, EC 2.7.11.1, Mammalian STE20-like protein kinase 3, MST-3, MST3serine/threonine kinase 24 (Ste20, yeast homolog), serine/threonine kinase 24, serine/threonine-protein kinase 24, STE20, STE20-like kinase 3, STE20-like kinase MST3, sterile 20-like kinase 3, STK3yeast) |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?