missing translation for 'onlineSavingsMsg'
Learn More

MST2/STK3 Rabbit anti-Human, Mouse, Rat, Clone: 5J1I4, Novus Biologicals™

Product Code. 18321626
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18321626 20 μg 20µL
18313563 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18321626 Supplier Novus Biologicals Supplier No. NBP31673720UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

MST2/STK3 Monoclonal antibody specifically detects MST2/STK3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen MST2/STK3
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 5J1I4
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias EC 2.7.11, FLJ90748, KRS1, Mammalian STE20-like protein kinase 2, MST-2, MST2EC 2.7.11.1, serine/threonine kinase 3, serine/threonine kinase 3 (STE20 homolog, yeast), serine/threonine kinase 3 (Ste20, yeast homolog), serine/threonine-protein kinase 3, Serine/threonine-protein kinase Krs-1, STE20-like kinase MST2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human MST2/STK3 (Q13188).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 6788
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.