missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MST2/STK3 Antibody (3C3), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26298-100ul
This item is not returnable.
View return policy
Description
MST2/STK3 Monoclonal antibody specifically detects MST2/STK3 in Human samples. It is validated for Western Blot, ELISA
Specifications
| MST2/STK3 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| 3C3 | |
| Western Blot 1:500-1:2000, ELISA | |
| EC 2.7.11, FLJ90748, KRS1, Mammalian STE20-like protein kinase 2, MST-2, MST2EC 2.7.11.1, serine/threonine kinase 3, serine/threonine kinase 3 (STE20 homolog, yeast), serine/threonine kinase 3 (Ste20, yeast homolog), serine/threonine-protein kinase 3, Serine/threonine-protein kinase Krs-1, STE20-like kinase MST2 | |
| A synthesized peptide derived from Human MST2/STK3 [UniProt Q13188] | |
| 100 μL | |
| Apoptosis | |
| 6788 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction