missing translation for 'onlineSavingsMsg'
Learn More

MST2/STK3 Antibody (1B3), Novus Biologicals™

Product Code. 18362139 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18362139 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18362139 Supplier Novus Biologicals Supplier No. H00006788M11

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MST2/STK3 Monoclonal antibody specifically detects MST2/STK3 in Human, Mouse samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen MST2/STK3
Applications Western Blot, ELISA
Classification Monoclonal
Clone 1B3
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_006272
Gene Alias EC 2.7.11, FLJ90748, KRS1, Mammalian STE20-like protein kinase 2, MST-2, MST2EC 2.7.11.1, serine/threonine kinase 3, serine/threonine kinase 3 (STE20 homolog, yeast), serine/threonine kinase 3 (Ste20, yeast homolog), serine/threonine-protein kinase 3, Serine/threonine-protein kinase Krs-1, STE20-like kinase MST2
Host Species Mouse
Immunogen STK3 (NP_006272.1, 253 a.a. ~ 350 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. DFVKKCLVKNPEQRATATQLLQHPFIKNAKPVSILRDLITEAMEIKAKRHEEQQRELEEEEENSDEDELDSHTMVKTSVESVGTMRATSTMSEGAQTM
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 6788
Target Species Human, Mouse
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.