missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MST1/STK4 Antibody (07), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-06389-100ul
This item is not returnable.
View return policy
Description
MST1/STK4 Monoclonal antibody specifically detects MST1/STK4 in Human samples. It is validated for ELISA, Sandwich ELISA Capture
Specifications
| MST1/STK4 | |
| Monoclonal | |
| Unconjugated | |
| AI447339;AU020804;Kas-2;KRS2;MST1;STK4;YSK3 | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human MST1/STK4 (Uniprot#: Q13043-1; Glu2-Phe487). | |
| 100 μg | |
| Apoptosis, Autophagy, Cardiovascular Biology, Cell Biology, Signal Transduction | |
| Human | |
| Purified |
| ELISA, Sandwich ELISA Capture | |
| 07 | |
| 0.2 um filtered solution in PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction