Learn More
MST1 (STK4) Polyclonal Antibody
GREENER_CHOICE
MST1 (STK4) Polyclonal Antibody
Brand:  Invitrogen PA596907
Product Code. 16559741
  • £439.00 / 100µL
Description
MST1 (STK4) Polyclonal Antibody for Western Blot, IHC (P), IP
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MST1 (STK4)
Polyclonal
Unconjugated
Stk4
AI447339; AU020804; EC 2.7.11.1; Kas-2; kinase MST1; kinase responsive to stress 2; KRS2; Mammalian STE20-like protein kinase 1; mammalian sterile 20-like 1; MST1; MST-1; MST1/C; MST1/N; serine/threonine kinase 4; serine/threonine-protein kinase 4; Serine/threonine-protein kinase 4 18kDa subunit; Serine/threonine-protein kinase 4 37kDa subunit; Serine/threonine-protein kinase Krs-2; STE20-like kinase MST1; sterile 20-like kinase 1; STK4; TIIAC; Yeast Sps1/Ste20-related kinase 3; Ysk3
Rabbit
Affinity Chromatography
RUO
311622, 58231, 6789
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q13043, Q9JI11
Stk4
Recombinant fusion protein containing a sequence corresponding to amino acids 303-487 of human MST1/STK4 (NP_0062731).
100 μL
Primary
Human, Mouse, Rat