Learn More
MSSA Multi-Stream Solution Analyzer
Product Code. 31783687 Shop All Applied Biosystems Products
Change view
Click to view available options
Type:
MSSA Multi-Stream Solution Analyzer
Packungsgröße:
Each
Artikelnummer. Type Packungsgröße
31783687 MSSA Multi-Stream Solution Analyzer Each
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 31783687 Lieferant ABI Applied Biosystems Lieferanten-Nr. 19506
 Restricted Item
Estimated Shipment: 22-12-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
missing translation for 'validMainframeMsg'
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Thermo Scientific™ MSSA provides a continuous window into the hydrometallurgical process which allows operators or an automatic control system to increase plant efficiency, quality and recovery.
Thermo Scientific™ MSSA provides a continuous window into the hydrometallurgical process which allows operators or an automatic control system to increase plant efficiency, quality and recovery. The Thermo Scientific Multi-Stream Solution Analyzer, MSSA, provides continuous online analysis of solution streams and is capable ofmeasuring elements as low as sulphur in the periodic table. This provides costeffective, real-time analysis of key elements and other variables from sampledstream lines in hydrometallurgical operations such as SX/EW and refineries.
Key Features
  • Simultaneously measures measure 8 elements
  • Receives between 1 and 16 streams per analyzer
  • Energy Dispersive XRF technique of analysis
  • Stainless-steel frame and construction
  • Stainless-steel or polypropylene wetted parts
  • Simple, low maintenance design
  • De-multiplexing of streams available if required
  • Metallurgical shift sampling available
  • Flush capabilities for scaling solutions
Specific Benefits
  • Monitors the transfer of metal species through different stages of the plant, viz: pregnant liquor solution, organic and electrolytes.
  • Monitors contaminant species in copper SX/EW in aqueous streams
  • Automated rectifiers in the EW plant
  • Optimized electrical energy use in the EW plant by minimizing peak power loads
  • Optimized current density based on metal concentration in the electrolyte to improve cathode quality
  • Automated reagent dosing
  • Manpower savings due to the decrease in control samples analyzed by the laboratory
Ordering Alerts:
A single-stream analyzer (SSSA) is also available; for information contact your Thermo Scientific Sales Representative.
TRUSTED_SUSTAINABILITY
Spezifikation
Type MSSA Multi-Stream Solution Analyzer
Description Multi-Stream Solution Analyzer, SamStat-30 Slurry Sampling Station (For details contact your Sales Representative)
Certifications/Compliance Manufactured in an ISO 9001:2000 certified facility.
Name des Produkts
Wählen Sie ein Thema
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.