missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MSRB3 Antibody (S03-9C6), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19701-100ul
This item is not returnable.
View return policy
Description
MSRB3 Monoclonal antibody specifically detects MSRB3 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| MSRB3 | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| S03-9C6 | |
| Unconjugated | |
| DFNB74, DKFZp686C1178, EC 1.8.4.-, EC 1.8.4.12, FLJ36866, methionine sulfoxide reductase B3, methionine-R-sulfoxide reductase B3, methionine-R-sulfoxide reductase B3, mitochondrial, MsrB3 | |
| Recombinant protein of human MSRB3 (Uniprot # Q8IXL7) | |
| 100 μg | |
| metabolism | |
| 253827 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction