missing translation for 'onlineSavingsMsg'
Learn More

MSRB3 Antibody - BSA Free, R&D Systems™

Product Code. 18611471 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.02 mL
0.1 mL
Unit Size:
0.01mL
0.02mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18611471 0.1 mL 0.01mL
18611381 0.02 mL 0.02mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18611471 Supplier Novus Biologicals Supplier No. NBP2936940.1ml
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody

MSRB3 Polyclonal specifically detects MSRB3 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry/Immunofluorescence, Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MSRB3
Applications Immunocytochemistry/Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200
Formulation PBS (pH 7.3), 50% glycerol
Gene Alias DFNB74, DKFZp686C1178, EC 1.8.4.-, EC 1.8.4.12, FLJ36866, methionine sulfoxide reductase B3, methionine-R-sulfoxide reductase B3, methionine-R-sulfoxide reductase B3, mitochondrial, MsrB3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-185 of human MSRB3 (NP_001026849.1). MSAFNLLHLVTKSQPVALRACGLPSGSCRDKKNCKVVFSQQELRKRLTPLQYHVTQEKGTESAFEGEYTHHKDPGIYKCVVCGTPLFKSETKFDSGSGWPSFHDVINSEAITFTDDFSYGMHRVETSCSQCGAHLGHIFDDGPRPTGKRYCINSAALSFTPADSSGTAEGGSGVASPAQADKAEL
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline metabolism
Primary or Secondary Primary
Gene ID (Entrez) 253827
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.