Learn More
Invitrogen™ MSRB2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5117128
Description
Positive Samples: Mouse heart, Mouse kidney, Mouse brain, Rat liver Immunogen sequence: GLTLGTAPRR AVRGQAGGGG PGTGPGLGEA GSLATCELPL AKSEWQKKLT PEQFYVTREK GTEPPFSGIY LNNKEAGMYH CVCCDSPLFS SEKKYCSGTG WPSFSEAHGT SGSDESHTGI LRRLDTSLGS ARTEVVCKQC EAHLGHVFPD GPGPNGQRFC INSVALKFKP RKH.
MSRB2 catalyzes the reduction of free and protein-bound methionine sulfoxide to methionine.
Specifications
| MSRB2 | |
| Polyclonal | |
| Unconjugated | |
| MSRB2 | |
| 2310050L06Rik; CBS1; CBS-1; CGI-131; methionine sulfoxide reductase B2; methionine-R-sulfoxide reductase B2, mitochondrial; Mrsb; Msrb; MsrB2; PILB; pilin-like transcription factor | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 22921, 361286, 76467 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 1.22 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q4FZX5, Q78J03, Q9Y3D2 | |
| MSRB2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 10-182 of human MSRB2 (NP_0363603). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.