missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MSRB2, Mouse anti-Human, Clone: 3F12, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00022921-M03.100ug
This item is not returnable.
View return policy
Description
Sequence: KEAGMYHCVCCDSPLFSSEKKYCSGTGWPSFSEAHGTSGSDESHTGILRRLDTSLGSARTEVVCKQCEAHLGHVFPDGPGPNGQRFCINSVALKFKPRKHSpecifications
| MSRB2 | |
| Monoclonal | |
| Unconjugated | |
| methionine sulfoxide reductase B2 | |
| CBS-1/CBS1/CGI-131/MGC26104/MSRB/PILB | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 3F12 | |
| In 1x PBS, pH 7.4 | |
| NM_012228 | |
| MSRB2 | |
| MSRB2 (NP_036360, 102 a.a. ∼ 201 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 22921 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction