missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MSRA Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14547-1-AP-150UL
This item is not returnable.
View return policy
Description
Methionine sulfoxide reductase (MsrA) reduces methionine sulfoxide (MetO) residues in proteins and free MetO to Methionine (Met). The catalytic activity of MsrA is dependent of bound metal and cofactors but it requires reducing equivalents from either DTT or a thioredoxin-regenerating system. MsrA plays an essential role in protecting cells against oxidative damage. The substrates of MsrA include calmodulin, HIV protease and a 1-proteinase-inhibitor. Recent studies indicate that there is a connection between MsrA and Alzheimer′s disease in mammals.Specifications
| MSRA | |
| Polyclonal | |
| Unconjugated | |
| MSRA | |
| MSRA, Peptide Met(O) reductase, PMSR | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 110265, 29447, 4482 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q923M1, Q9D6Y7, Q9UJ68 | |
| MSRA | |
| MSRA Fusion Protein Ag6053 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction