missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
MSPR/Ron Monoclonal antibody specifically detects MSPR/Ron in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | MSPR/Ron |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | CL0463 |
| Concentration | LYOPH |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1 μg/mL |
| Formulation | Lyophilized in 5% Trehalose. |
| Gene Alias | CD136, CD136 antigen, CDw136c-met-related tyrosine kinase, EC 2.7.10, EC 2.7.10.1, macrophage stimulating 1 receptor (c-met-related tyrosine kinase), MSP receptor, p185-Ron, Protein-tyrosine kinase 8, PTK8, PTK8 protein tyrosine kinase 8, RONmacrophage-stimulating protein receptor, soluble RON variant 1, soluble RON variant 2, soluble RON variant 3, soluble RON variant 4 |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?